Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RS-102221 hydrochloride | 187397-18-8 | 98.0% | 649.08 g/mol | C27H32ClF3N4O7S | 50 MG
SDP

Catalog No. 5000211997
Encompass_Preferred

RS-102221 hydrochloride is a selective 5-HT2C receptor antagonist used in preclinical research to probe serotonin 5-HT2C-mediated signaling. It exhibits high affinity (Ki ≈ 10 nM) and strong selectivity versus related receptor subtypes and is supplied as a white to off-white solid for laboratory use.

  • Selective 5-HT2C receptor antagonist with high affinity.
  • Approximately 100-fold selectivity over 5-HT2A and 5-HT2B receptors.
  • Supplied as a white to off-white solid suitable for research.
  • Available in multiple small-scale sizes for laboratory experiments.
  • Stable when stored under recommended conditions for powder and solutions.

Catalog No. 50-002-11997 Supplier Medchemexpress LLC Supplier No. HY101365A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RS-102221 hydrochloride is a selective 5-HT2C receptor antagonist used in preclinical research to probe serotonin 5-HT2C-mediated signaling. It exhibits high affinity (Ki ≈ 10 nM) and strong selectivity versus related receptor subtypes and is supplied as a white to off-white solid for laboratory use.

  • Selective 5-HT2C receptor antagonist with high affinity.
  • Approximately 100-fold selectivity over 5-HT2A and 5-HT2B receptors.
  • Supplied as a white to off-white solid suitable for research.
  • Available in multiple small-scale sizes for laboratory experiments.
  • Stable when stored under recommended conditions for powder and solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.