Learn More
Abcam RS 102895 hydrochloride, CCR2 receptor antagonist, 50MG
Catalog No. 502707236
Encompass_Preferred
MW 426.9 Da, Purity >99%. Selective CCR2 receptor antagonist (IC₅₀ values are 360 nM and 17.8 μM at CCR2 and CCR1 receptors, respectively). Inhibits MCP-1 and MCP-3 stimulated calcium influx (IC₅₀ values are 32 and 130 nM, respectively). Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7236 Supplier Abcam Supplier No. AB12081250MG
MW 426.9 Da, Purity >99%. Selective CCR2 receptor antagonist (IC₅₀ values are 360 nM and 17.8 μM at CCR2 and CCR1 receptors, respectively). Inhibits MCP-1 and MCP-3 stimulated calcium influx (IC₅₀ values are 32 and 130 nM, respectively). Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement