Learn More
Cayman Chemical RS 102895 (hydrochloride), Purity: ≥98%, CAS: 1173022-16-6, 10 mg
SDP
Catalog No. 502968032
Encompass_Preferred
A chemokine receptor CCR2 antagonist (IC50 = 0.36 µM); inhibits the related CCR1 receptor with an IC50 value of 17.8 µM and inhibits adrenergic α1a, α1d, and 5HT1A receptors with IC50 values of 0.13, 0.32, and 47 µM, respectively
Catalog No. 50-296-8032 Supplier Cayman Chemical Supplier No. 1820710 MG
A chemokine receptor CCR2 antagonist (IC50 = 0.36 µM); inhibits the related CCR1 receptor with an IC50 value of 17.8 µM and inhibits adrenergic α1a, α1d, and 5HT1A receptors with IC50 values of 0.13, 0.32, and 47 µM, respectively