Learn More
Selleck Chemical LLC RS-127445 S2698-50mg
SDP
Supplier:  Selleck Chemical LLC S269850MG
Encompass_Preferred
RS-127445 (MT500) is a selective 5-HT2B receptor antagonist with pKi of 9 5 and pIC50 of 10 4 exhibits 1000-fold selectivity against other 5-HT receptors
Catalog No. 50-311-2352