Learn More
Medchemexpress LLC RS-127445 | 199864-87-4 | C17H16FN3 | 1 ML
SDP
Catalog No. 5000376619
Encompass_Preferred
RS-127445 is a selective, high affinity, and orally bioavailable 5-HT2B receptor antagonist. It demonstrates a pKi of 9.5 and exhibits over 1000-fold selectivity for this receptor compared to numerous other receptor and ion channel binding sites.
  • Selective 5-HT2B receptor antagonist
  • High affinity (pKi of 9.5)
  • Orally bioavailable
  • Over 1000-fold selectivity compared to other receptors
  • Potently blocks 5-HT evoked increases in intracellular calcium concentrations
Catalog No. 50-003-76619 Supplier Medchemexpress LLC Supplier No. HY15419A10MM/1ML
RS-127445 is a selective, high affinity, and orally bioavailable 5-HT2B receptor antagonist. It demonstrates a pKi of 9.5 and exhibits over 1000-fold selectivity for this receptor compared to numerous other receptor and ion channel binding sites.
  • Selective 5-HT2B receptor antagonist
  • High affinity (pKi of 9.5)
  • Orally bioavailable
  • Over 1000-fold selectivity compared to other receptors
  • Potently blocks 5-HT evoked increases in intracellular calcium concentrations