Learn More
Medchemexpress LLC RS-127445 | 199864-87-4 | C17H16FN3 | 5 MG
SDP
Catalog No. 5000376355
Encompass_Preferred
This compound is a selective, high-affinity, and orally bioavailable antagonist of the 5-HT2B receptor, exhibiting a pKi of 9.5. It demonstrates significantly higher selectivity (1000-fold) for this receptor compared to numerous other receptor and ion channel binding sites.
  • High affinity for the target receptor.
  • Orally bioavailable.
  • Shows high selectivity, reducing off-target effects.
  • Effective in inhibiting serotonin-induced intracellular calcium increases.
  • Proven antagonist activity in cellular assays.
Catalog No. 50-003-76355 Supplier Medchemexpress LLC Supplier No. HY15419A5MG
This compound is a selective, high-affinity, and orally bioavailable antagonist of the 5-HT2B receptor, exhibiting a pKi of 9.5. It demonstrates significantly higher selectivity (1000-fold) for this receptor compared to numerous other receptor and ion channel binding sites.
  • High affinity for the target receptor.
  • Orally bioavailable.
  • Shows high selectivity, reducing off-target effects.
  • Effective in inhibiting serotonin-induced intracellular calcium increases.
  • Proven antagonist activity in cellular assays.