Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RS-127445 | 199864-87-4 | C17H16FN3 | 50 MG
SDP

Catalog No. 5000376136
Encompass_Preferred

RS-127445 is a selective, high-affinity, orally bioavailable 5-HT2B receptor antagonist. It demonstrates 1000-fold selectivity for this receptor compared to numerous other receptor and ion channel binding sites, making it suitable for research applications.

  • High affinity for the 5-HT2B receptor (pKi = 9.5)
  • 1000-fold selectivity for 5-HT2B receptor
  • Orally bioavailable
  • Appearance: solid, white to off-white
  • Purity: 99.95%
  • Store at 4°C, sealed, away from moisture; In solvent: -80°C for 6 months, -20°C for 1 month (sealed, away from moisture)

Catalog No. 50-003-76136 Supplier Medchemexpress LLC Supplier No. HY15419A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RS-127445 is a selective, high-affinity, orally bioavailable 5-HT2B receptor antagonist. It demonstrates 1000-fold selectivity for this receptor compared to numerous other receptor and ion channel binding sites, making it suitable for research applications.

  • High affinity for the 5-HT2B receptor (pKi = 9.5)
  • 1000-fold selectivity for 5-HT2B receptor
  • Orally bioavailable
  • Appearance: solid, white to off-white
  • Purity: 99.95%
  • Store at 4°C, sealed, away from moisture; In solvent: -80°C for 6 months, -20°C for 1 month (sealed, away from moisture)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.