Learn More
Selleck Chemical LLC RS-127445 5mg 199864-87-4 MT500
SDP
Supplier:  Selleck Chemical LLC S26985MG
Encompass_Preferred
RS-127445 (MT500) is a selective 5-HT2B receptor antagonist with pKi of 9.5 and pIC50 of 10.4, exhibits >1000-fold selectivity against other 5-HT receptors. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2799