Learn More
Medchemexpress LLC Rs-127445 hydrochloride | 199864-86-3 | 99.8% | 317.79 g·mol⁻¹ | C17H17ClFN3 | 10 MG
SDP
Catalog No. 5000287446
Encompass_Preferred
RS-127445 hydrochloride is a selective, high-affinity 5-HT2B receptor antagonist (pKi ≈ 9.5) provided as a research-grade chemical. It shows approximately 1,000-fold selectivity versus other receptors and is supplied as a solid (powder) or as a ready-to-use 10 mM solution in DMSO. Intended for research use only.
Catalog No. 50-002-87446 Supplier Medchemexpress LLC Supplier No. HY1541910MG
RS-127445 hydrochloride is a selective, high-affinity 5-HT2B receptor antagonist (pKi ≈ 9.5) provided as a research-grade chemical. It shows approximately 1,000-fold selectivity versus other receptors and is supplied as a solid (powder) or as a ready-to-use 10 mM solution in DMSO. Intended for research use only.