Learn More
Medchemexpress LLC RS-127445 hydrochloride | 199864-86-3 | C17H17ClFN3 | 100 MG
SDP
Catalog No. 5000376183
Encompass_Preferred
RS-127445 hydrochloride is a selective, high-affinity, and orally bioavailable 5-HT2B receptor antagonist. It exhibits 1000-fold selectivity for the 5-HT2B receptor compared to numerous other receptor and ion channel binding sites, making it a valuable tool for research into serotonin receptor activity and related biological pathways.
  • Selective and high-affinity 5-HT2B receptor antagonist
  • Orally bioavailable
  • Exhibits 1000-fold selectivity over many other receptors
  • Potently blocks 5-HT evoked increases in intracellular calcium concentrations
  • Reduces faecal output in rats dose-dependently
Catalog No. 50-003-76183 Supplier Medchemexpress LLC Supplier No. HY15419100MG
RS-127445 hydrochloride is a selective, high-affinity, and orally bioavailable 5-HT2B receptor antagonist. It exhibits 1000-fold selectivity for the 5-HT2B receptor compared to numerous other receptor and ion channel binding sites, making it a valuable tool for research into serotonin receptor activity and related biological pathways.
  • Selective and high-affinity 5-HT2B receptor antagonist
  • Orally bioavailable
  • Exhibits 1000-fold selectivity over many other receptors
  • Potently blocks 5-HT evoked increases in intracellular calcium concentrations
  • Reduces faecal output in rats dose-dependently