Learn More
Medchemexpress LLC RS-127445 hydrochloride | 199864-86-3 | C17H17ClFN3 | 50 MG
SDP
Catalog No. 5000376663
Encompass_Preferred
RS-127445 hydrochloride is a selective, high-affinity, orally bioavailable 5-HT2B receptor antagonist with a pKi of 9.5. It demonstrates 1000-fold selectivity for this receptor compared to numerous other receptor and ion channel binding sites. It is intended for research use only.
  • Selective 5-HT2B receptor antagonist
  • High-affinity binding
  • Orally bioavailable
  • Demonstrates 1000-fold selectivity
  • Suitable for research applications
Catalog No. 50-003-76663 Supplier Medchemexpress LLC Supplier No. HY1541950MG
RS-127445 hydrochloride is a selective, high-affinity, orally bioavailable 5-HT2B receptor antagonist with a pKi of 9.5. It demonstrates 1000-fold selectivity for this receptor compared to numerous other receptor and ion channel binding sites. It is intended for research use only.
  • Selective 5-HT2B receptor antagonist
  • High-affinity binding
  • Orally bioavailable
  • Demonstrates 1000-fold selectivity
  • Suitable for research applications