Learn More
TARGETMOL CHEMICALS INC RS 15385-198 5MG
SDP
Catalog No. 502576447
Encompass_Preferred
Also available in 1mg 10mg 25mg 50mg 100mg 500mg and bulk. Please contact Fisher for quotes. RS 15385-198 is a less active isomer of Delequamine. Delequamine is a selective alpha2-adrenergic receptor antagonist. purity: 98%
Catalog No. 50-257-6447 Supplier TARGETMOL CHEMICALS INC Supplier No. T68051L5MG
Also available in 1mg 10mg 25mg 50mg 100mg 500mg and bulk. Please contact Fisher for quotes. RS 15385-198 is a less active isomer of Delequamine. Delequamine is a selective alpha2-adrenergic receptor antagonist. purity: 98%