Learn More
Apexbio Technology LLC RS 16566 dihydrochloride 10mg
SDP
Catalog No. 5000091020
Encompass_Preferred
RS 16566 dihydrochloride is a small-molecule inhibitor targeting the receptor tyrosine kinase Tie2 It is designed to inhibit Tie2 thereby interrupting downstream Angiopoietin-mediated signaling pathways involved in vascular formation endothelial stability and tumor-associated vascular remodeling RS 16566 dihydrochloride exerts its biological activity primarily through prevention of Tie2 phosphorylation Based on these pharmacological properties RS 16566 dihydrochloride holds research potential in the investigation of angiogenesis and tumor progression within oncology and vascular biology studies
Catalog No. 50-000-91020 Supplier Apexbio Technology LLC Supplier No. B654810
RS 16566 dihydrochloride is a small-molecule inhibitor targeting the receptor tyrosine kinase Tie2 It is designed to inhibit Tie2 thereby interrupting downstream Angiopoietin-mediated signaling pathways involved in vascular formation endothelial stability and tumor-associated vascular remodeling RS 16566 dihydrochloride exerts its biological activity primarily through prevention of Tie2 phosphorylation Based on these pharmacological properties RS 16566 dihydrochloride holds research potential in the investigation of angiogenesis and tumor progression within oncology and vascular biology studies