Learn More
Medchemexpress LLC HY-101336 100mg Medchemexpress, RS 17053 hydrochloride CAS:169505-93-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY101336100MG
Encompass_Preferred
Medchemexpress, HY-101336 100mg RS 17053 hydrochloride CAS:169505-93-5 RS 17053 hydrochloride is a potent and selective α1 A adrenoceptor antagonist, with a pK i value of 9.1 in native cell membrane and a pA 2 value of 9.8 in functional assays. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2096