Learn More
Medchemexpress LLC Rs 17053 hydrochloride | 169505-93-5 | 99.9% | 449.41 g/mol | C24H30Cl2N2O2 | 10 MG
SDP
Catalog No. 5000211385
Encompass_Preferred
RS 17053 hydrochloride is a potent, selective α1A adrenoceptor antagonist for pharmacological research. It exhibits high affinity in native membranes (pKi 9.1) and functional potency (pA2 9.8), with marked selectivity versus α1B and α1D subtypes. The material is supplied as a high-purity solid and is provided in multiple sizes and a DMSO solution format for laboratory use.
  • Potent, selective α1A adrenoceptor antagonist with reported pKi 9.1 and pA2 9.8.
  • Substantial selectivity versus α1B and α1D receptor subtypes.
  • High purity suitable for sensitive pharmacology assays.
  • Available as a solid and as a ready-to-use DMSO solution for convenience.
  • Intended for research use in receptor binding and functional studies.
Catalog No. 50-002-11385 Supplier Medchemexpress LLC Supplier No. HY10133610MG
RS 17053 hydrochloride is a potent, selective α1A adrenoceptor antagonist for pharmacological research. It exhibits high affinity in native membranes (pKi 9.1) and functional potency (pA2 9.8), with marked selectivity versus α1B and α1D subtypes. The material is supplied as a high-purity solid and is provided in multiple sizes and a DMSO solution format for laboratory use.
  • Potent, selective α1A adrenoceptor antagonist with reported pKi 9.1 and pA2 9.8.
  • Substantial selectivity versus α1B and α1D receptor subtypes.
  • High purity suitable for sensitive pharmacology assays.
  • Available as a solid and as a ready-to-use DMSO solution for convenience.
  • Intended for research use in receptor binding and functional studies.