Learn More
Medchemexpress LLC Rs 17053 hydrochloride | 169505-93-5 | MFCD09971032 | 99.9% | 449.41 | C24H30Cl2N2O2 | 50 MG
SDP
Catalog No. 5000212515
Encompass_Preferred
RS 17053 hydrochloride is a research-grade small molecule that functions as a potent, selective α1A adrenoceptor antagonist for pharmacological studies. Supplied as a solid with high purity, it includes manufacturer-specified solubility, formulation, and storage guidance for both in vitro and in vivo use.
  • Potent, selective α1A adrenoceptor antagonist (pKi 9.1; pA2 9.8).
  • High purity: 99.9%.
  • Solid form, white to off-white.
  • Soluble in DMSO (~100 mg/mL); ultrasonic assistance recommended.
  • Compatible with multiple in vivo formulations at ≥2.08 mg/mL for clear solutions.
  • Stable with recommended storage: sealed at 4°C; in solution -80°C for long-term.
Catalog No. 50-002-12515 Supplier Medchemexpress LLC Supplier No. HY10133650MG
RS 17053 hydrochloride is a research-grade small molecule that functions as a potent, selective α1A adrenoceptor antagonist for pharmacological studies. Supplied as a solid with high purity, it includes manufacturer-specified solubility, formulation, and storage guidance for both in vitro and in vivo use.
  • Potent, selective α1A adrenoceptor antagonist (pKi 9.1; pA2 9.8).
  • High purity: 99.9%.
  • Solid form, white to off-white.
  • Soluble in DMSO (~100 mg/mL); ultrasonic assistance recommended.
  • Compatible with multiple in vivo formulations at ≥2.08 mg/mL for clear solutions.
  • Stable with recommended storage: sealed at 4°C; in solution -80°C for long-term.