Learn More
Heidolph™ RS 232 Cable (9-pole)
SDP
Catalog No. 036306014 Shop All Heidolph North America Products
Change view
Click to view available options
Includes:
Cable
Catalog No. Includes
03-630-6014 Cable
1 options
Catalog No. 03-630-6014 Supplier Heidolph™ Supplier No. 036306014
Add to Cart
Add to Cart
For use with Hei-VAP Ultimate Rotary Evaporator, Hei-PLATE Ultimate Magnetic Stirring Hotplate, Hei-TORQUE Ultimate Overhead Stirrer, and Hei-FLOW Ultimate Peristaltic Pump
RS 232 cable for connecting free Hei-Control software. Utilize the optional, free software to control and document the entire process. Connect up to four units at the same time
Specifications
Color Gray
Connection Type 9-Pole
Type RS232 Cable Connection
For Use With (Equipment) Hei-VAP Ultimate Rotary Evaporator, Hei-PLATE Ultimate Magnetic Stirring Hotplate, Hei-TORQUE Ultimate Overhead Stirrer, Hei-FLOW Ultimate Peristaltic Pump
Includes Cable
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov