Learn More
Medchemexpress LLC HY-112484 10mg Medchemexpress, RS-246204 CAS:878451-87-7 Purity:>98%
SDP
Catalog No. 501969098
Encompass_Preferred
Medchemexpress, HY-112484 10mg RS-246204 CAS:878451-87-7 RS-246204 is a R-spondin-1 substitute compound that is able initiate small intestinal organoids without the use of the R-spondin-1 protein. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9098 Supplier Medchemexpress LLC Supplier No. HY11248410MG
Medchemexpress, HY-112484 10mg RS-246204 CAS:878451-87-7 RS-246204 is a R-spondin-1 substitute compound that is able initiate small intestinal organoids without the use of the R-spondin-1 protein. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.