Learn More
Medchemexpress LLC RS-25344 hydrochloride | 152815-28-6 | 99.9% | 411.80 | 100 MG
SDP
Catalog No. 5000345416
Encompass_Preferred
RS-25344 hydrochloride is a selective cAMP-phosphodiesterase 4 (PDE 4; PDE IV) inhibitor with an IC50 of 0.28 nM in human lymphocytes. It exhibits weak inhibitory effects on PDE I, II, and III (IC50 of >100 μM, 160 μM, and 330 μM, respectively).
  • Has anti-inflammatory, memory- and cognition-enhancing, and antineoplastic effects
  • Potent inhibitor of sperm PDE activity with an effective concentration of 0.3 nM
  • Increases stomach weights and gastric retention in mice
Catalog No. 50-003-45416 Supplier Medchemexpress LLC Supplier No. HY108621100MG
RS-25344 hydrochloride is a selective cAMP-phosphodiesterase 4 (PDE 4; PDE IV) inhibitor with an IC50 of 0.28 nM in human lymphocytes. It exhibits weak inhibitory effects on PDE I, II, and III (IC50 of >100 μM, 160 μM, and 330 μM, respectively).
  • Has anti-inflammatory, memory- and cognition-enhancing, and antineoplastic effects
  • Potent inhibitor of sperm PDE activity with an effective concentration of 0.3 nM
  • Increases stomach weights and gastric retention in mice