Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RS-25344 hydrochloride | 152815-28-6 | MFCD16618380 | 99.9% | 411.80 g·mol^-1 | C19H14ClN5O4 | 5 MG
SDP

Catalog No. 5000263406
Encompass_Preferred

RS-25344 hydrochloride is the hydrochloride salt of RS-25344, a potent and selective cAMP-phosphodiesterase 4 (PDE4) inhibitor used in biochemical and pharmacological research. CAS 152815-28-6; molecular formula C19H14ClN5O4; molecular weight 411.80 g·mol⁻1. Supplied as a white to off-white solid or as DMSO solution for in vitro studies.

  • Potent PDE4 inhibition with IC50 0.28 nM in human lymphocytes.
  • High chemical purity (99.93%) suitable for research applications.
  • Soluble in DMSO (≈83.33 mg/mL); ultrasonic agitation may be required.
  • White to off-white solid appearance for easy handling and weighing.
  • Storage: solid at 4°C; solutions -80°C (6 months) or -20°C (1 month).
  • Reported biological activities include anti-inflammatory and cognition-enhancing effects.

Catalog No. 50-002-63406 Supplier Medchemexpress LLC Supplier No. HY1086215MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RS-25344 hydrochloride is the hydrochloride salt of RS-25344, a potent and selective cAMP-phosphodiesterase 4 (PDE4) inhibitor used in biochemical and pharmacological research. CAS 152815-28-6; molecular formula C19H14ClN5O4; molecular weight 411.80 g·mol⁻1. Supplied as a white to off-white solid or as DMSO solution for in vitro studies.

  • Potent PDE4 inhibition with IC50 0.28 nM in human lymphocytes.
  • High chemical purity (99.93%) suitable for research applications.
  • Soluble in DMSO (≈83.33 mg/mL); ultrasonic agitation may be required.
  • White to off-white solid appearance for easy handling and weighing.
  • Storage: solid at 4°C; solutions -80°C (6 months) or -20°C (1 month).
  • Reported biological activities include anti-inflammatory and cognition-enhancing effects.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.