Learn More
Cayman Chemical RS 25344 (hydrochloride), Purity: ≥95%, CAS: 152815-28-6, 100 mg
SDP
Catalog No. 502917788
Encompass_Preferred
A PDE4 inhibitor (IC50 = 0.3 nM); selective for PDE4 over PDE1, -2, and -3 (IC50s = >100 µM for all); inhibits concanavalin A-induced IL-5 release and LPS-induced TNF-α release in isolated human PBMCs (EC50s = 0.3 and 5.4 nM, respectively)
Catalog No. 50-291-7788 Supplier Cayman Chemical Supplier No. 37793100 MG
A PDE4 inhibitor (IC50 = 0.3 nM); selective for PDE4 over PDE1, -2, and -3 (IC50s = >100 µM for all); inhibits concanavalin A-induced IL-5 release and LPS-induced TNF-α release in isolated human PBMCs (EC50s = 0.3 and 5.4 nM, respectively)