Learn More
Selleck Chemical LLC (R)-Nepicastat HCl S4926-10mg
SDP
Catalog No. 503111932
Encompass_Preferred
(R)-Nepicastat HCl (RS-25560-198) the R-enantiomer of Nepicastat HCl is a potent and selective inhibitor with IC50 of 25 1 nM and 18 3 nM for bovine and human dopamine- -hydroxylase with negligible affinity for twelve other enzymes and thirteen neurotransmitter receptors
Catalog No. 50-311-1932 Supplier Selleck Chemical LLC Supplier No. S492610MG
(R)-Nepicastat HCl (RS-25560-198) the R-enantiomer of Nepicastat HCl is a potent and selective inhibitor with IC50 of 25 1 nM and 18 3 nM for bovine and human dopamine- -hydroxylase with negligible affinity for twelve other enzymes and thirteen neurotransmitter receptors