Learn More
Medchemexpress LLC HY-15418 50mg , RS 504393 CAS:300816-15-3 Purity:98%
SDP
Catalog No. 501493940
Encompass_Preferred
Medchemexpress, HY-15418 50mg RS 504393 CAS:300816-15-3 Formula:C25H27N3O3 IC50: 98 nM (CCR2b) Purity:98% RS 504393 is a selective CCR2 chemokine receptor antagonist ( IC 50 values are 98 nM and > 100 μM for inhibition of human recombinant CCR2b and CCR1 receptors respectively). Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3940 Supplier Medchemexpress LLC Supplier No. HY1541850MG
Medchemexpress, HY-15418 50mg RS 504393 CAS:300816-15-3 Formula:C25H27N3O3 IC50: 98 nM (CCR2b) Purity:98% RS 504393 is a selective CCR2 chemokine receptor antagonist ( IC 50 values are 98 nM and > 100 μM for inhibition of human recombinant CCR2b and CCR1 receptors respectively). Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.