Learn More
TARGETMOL CHEMICALS INC RS 504393 10MG
SDP
Catalog No. 502579780
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RS 504393 is a highly selective CCR2 chemokine receptor antagonist (IC50s 89 nM and > 100 uM for human recombinant CCR2 and CCR1). purity: 98%
Catalog No. 50-257-9780 Supplier TARGETMOL CHEMICALS INC Supplier No. T538410MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RS 504393 is a highly selective CCR2 chemokine receptor antagonist (IC50s 89 nM and > 100 uM for human recombinant CCR2 and CCR1). purity: 98%