Learn More
Medchemexpress LLC HY-15418 25mg , RS 504393 CAS:300816-15-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1541825MG
Encompass_Preferred
HY-15418 25mg RS 504393 CAS: 300816-15-3 RS 504393 is a selective CCR2 chemokine receptor antagonist (IC50 values are 89 nM and > 100 μM for inhibition of human recombinant CCR2 and CCR1 receptors respectively). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0909