Learn More
Abcam RS 504393, CCR2 chemokine receptor antagonist, 10MG
Catalog No. 502707235
Encompass_Preferred
MW 417.5 Da, Purity >98%. Highly selective CCR2 chemokine receptor antagonist (IC₅₀ = 89 nM). Shows 700-fold greater binding for CCR2 vs. CCR1. Inhibits MCP-1 chemotaxis (IC₅₀ = 330 nM) and ischemia-reperfusion injury in kidneys.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7235 Supplier Abcam Supplier No. AB12081310MG
MW 417.5 Da, Purity >98%. Highly selective CCR2 chemokine receptor antagonist (IC₅₀ = 89 nM). Shows 700-fold greater binding for CCR2 vs. CCR1. Inhibits MCP-1 chemotaxis (IC₅₀ = 330 nM) and ischemia-reperfusion injury in kidneys.
The product is subject to the following: Abcam Restricted Use Statement