Learn More
Medchemexpress LLC Rs 67333 hydrochloride | 168986-60-5 | 99.7% | 389.36 | C19H30Cl2N2O2 | 100 MG
SDP
Catalog No. 5000213147
Encompass_Preferred
RS 67333 hydrochloride is a research-grade 5-HT4 (serotonin 4) receptor partial agonist supplied as the hydrochloride salt for use in neuroscience and pharmacology studies. It is commonly used to probe 5-HT4 receptor function in vitro and in vivo, with reported neuroprotective and pro-cognitive activity.
  • Partial agonist at 5-HT4 receptors, useful for receptor pharmacology studies.
  • Hydrochloride salt form for improved solubility in aqueous media.
  • High purity (99.68%) suitable for research applications.
  • Characterized by CAS 168986-60-5 and defined molecular properties for formulation.
  • Available in small research quantities (e.g., 100 MG) for in vitro and in vivo use.
  • Relevant for studies of cognition, neuroprotection, and serotonergic signaling.
Catalog No. 50-002-13147 Supplier Medchemexpress LLC Supplier No. HY101341100MG
RS 67333 hydrochloride is a research-grade 5-HT4 (serotonin 4) receptor partial agonist supplied as the hydrochloride salt for use in neuroscience and pharmacology studies. It is commonly used to probe 5-HT4 receptor function in vitro and in vivo, with reported neuroprotective and pro-cognitive activity.
  • Partial agonist at 5-HT4 receptors, useful for receptor pharmacology studies.
  • Hydrochloride salt form for improved solubility in aqueous media.
  • High purity (99.68%) suitable for research applications.
  • Characterized by CAS 168986-60-5 and defined molecular properties for formulation.
  • Available in small research quantities (e.g., 100 MG) for in vitro and in vivo use.
  • Relevant for studies of cognition, neuroprotection, and serotonergic signaling.