Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rs 67333 hydrochloride | 168986-60-5 | 99.7% | 389.36 g·mol⁻1 | C19H30Cl2N2O2 | 10 MG
SDP

Catalog No. 5000213172
Encompass_Preferred

RS 67333 hydrochloride is a research-grade small molecule that acts as a potent and selective partial agonist of the 5-HT4 (serotonin) receptor. It is commonly used in neuroscience research, including studies of neuroprotection and Alzheimer's disease models, to probe 5-HT4-mediated signaling.

  • Potent and selective 5-HT4 receptor partial agonist (pKi ≈8.7).
  • Used in neuroscience and neurodegeneration research models.
  • High purity (~99.7%).
  • Molecular weight 389.36 g·mol⁻1.
  • CAS number 168986-60-5.
  • Available in small research quantities, such as 10 MG, and stored sealed away from moisture.

Catalog No. 50-002-13172 Supplier Medchemexpress LLC Supplier No. HY10134110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RS 67333 hydrochloride is a research-grade small molecule that acts as a potent and selective partial agonist of the 5-HT4 (serotonin) receptor. It is commonly used in neuroscience research, including studies of neuroprotection and Alzheimer's disease models, to probe 5-HT4-mediated signaling.

  • Potent and selective 5-HT4 receptor partial agonist (pKi ≈8.7).
  • Used in neuroscience and neurodegeneration research models.
  • High purity (~99.7%).
  • Molecular weight 389.36 g·mol⁻1.
  • CAS number 168986-60-5.
  • Available in small research quantities, such as 10 MG, and stored sealed away from moisture.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.