Learn More
Medchemexpress LLC RS 67333 hydrochloride | 168986-60-5 | 99.7% | 389.36 g/mol | C19H30Cl2N2O2 | 5 MG
SDP
Catalog No. 5000213017
Encompass_Preferred
RS 67333 hydrochloride is a research-grade small-molecule partial agonist of the 5-HT4 (serotonin) receptor, supplied as a solid for laboratory use. It is intended for neuroscience and receptor-signaling studies and is provided with specification details and storage recommendations for research environments.
  • Potent, selective 5-HT4 receptor partial agonist.
  • High purity (≈99.7%).
  • Molecular weight 389.36 g/mol.
  • Solid, light yellow to yellow appearance.
  • Recommended for research use only with sealed, moisture-free storage.
Catalog No. 50-002-13017 Supplier Medchemexpress LLC Supplier No. HY1013415MG
RS 67333 hydrochloride is a research-grade small-molecule partial agonist of the 5-HT4 (serotonin) receptor, supplied as a solid for laboratory use. It is intended for neuroscience and receptor-signaling studies and is provided with specification details and storage recommendations for research environments.
  • Potent, selective 5-HT4 receptor partial agonist.
  • High purity (≈99.7%).
  • Molecular weight 389.36 g/mol.
  • Solid, light yellow to yellow appearance.
  • Recommended for research use only with sealed, moisture-free storage.