Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RS 67333 (hydrochloride) | 168986-60-5 | MFCD00938576 | 99.7% | 389.36 | C19H30Cl2N2O2 | 25 MG
SDP

Catalog No. 5000213145
Encompass_Preferred

RS 67333 hydrochloride is a research-grade, selective 5-HT4 receptor partial agonist used in preclinical and in vitro neuroscience studies to investigate serotonergic signaling and neuroprotective mechanisms.

  • Potent, selective 5-HT4 receptor partial agonist for neuroscience research.
  • High purity, 99.7% (as listed by manufacturer).
  • Molecular weight 389.36 g/mol; formula C19H30Cl2N2O2.
  • Solid, light yellow to yellow appearance supplied as a dry powder.
  • Storage: 4°C, sealed and away from moisture; in solvent store at -80°C for long-term.
  • Available as a 25 MG vial for laboratory use.
  • CAS number 168986-60-5.

Catalog No. 50-002-13145 Supplier Medchemexpress LLC Supplier No. HY10134125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RS 67333 hydrochloride is a research-grade, selective 5-HT4 receptor partial agonist used in preclinical and in vitro neuroscience studies to investigate serotonergic signaling and neuroprotective mechanisms.

  • Potent, selective 5-HT4 receptor partial agonist for neuroscience research.
  • High purity, 99.7% (as listed by manufacturer).
  • Molecular weight 389.36 g/mol; formula C19H30Cl2N2O2.
  • Solid, light yellow to yellow appearance supplied as a dry powder.
  • Storage: 4°C, sealed and away from moisture; in solvent store at -80°C for long-term.
  • Available as a 25 MG vial for laboratory use.
  • CAS number 168986-60-5.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.