Learn More
Medchemexpress LLC RS 67333 hydrochloride | 168986-60-5 | MFCD00938576 | 99.7% | 389.36 g/mol | C19H30Cl2N2O2 | 1 ML
SDP
Catalog No. 5000213018
Encompass_Preferred
RS 67333 hydrochloride is a research reagent that functions as a potent, selective partial agonist of the 5-HT4 (serotonin 4) receptor. It is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and in multiple solid pack sizes for use in pharmacology and neuroscience studies, including neuroprotection and Alzheimer's disease research. CAS: 168986-60-5; purity ~99.7%.
  • Potent, selective 5-HT4 receptor partial agonist.
  • Offered as 10 mM solution in DMSO (1 mL) and multiple solid sizes.
  • High listed purity (~99.7%).
  • Suitable for pharmacology and neuroscience research applications.
  • Provided with datasheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-13018 Supplier Medchemexpress LLC Supplier No. HY10134110MM 1ML
RS 67333 hydrochloride is a research reagent that functions as a potent, selective partial agonist of the 5-HT4 (serotonin 4) receptor. It is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and in multiple solid pack sizes for use in pharmacology and neuroscience studies, including neuroprotection and Alzheimer's disease research. CAS: 168986-60-5; purity ~99.7%.
  • Potent, selective 5-HT4 receptor partial agonist.
  • Offered as 10 mM solution in DMSO (1 mL) and multiple solid sizes.
  • High listed purity (~99.7%).
  • Suitable for pharmacology and neuroscience research applications.
  • Provided with datasheet, certificate of analysis, and safety data sheet.