Learn More
TARGETMOL CHEMICALS INC RS 67333 HYDROCHLORIDE 5MG
SDP
Catalog No. 502580885
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RS 67333 hydrochloride (RO5203648) is a potent and selective 5-HT4 receptor (5-HT4R) partial agonist with a pKi of 8.7 in guinea-pig striatum. RS 67333 hydrochloride exhibits lower affinities at several other receptors including 5-HT1A 5-HT1D 5-HT2A 5-HT2C dopamine D1 D2 and muscarinic M1-M3 receptors. RS 67333 hydrochloride shows neuroprotective effects and can be used for Alzheimer's disease research. purity: 98%
Catalog No. 50-258-0885 Supplier TARGETMOL CHEMICALS INC Supplier No. T232655MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RS 67333 hydrochloride (RO5203648) is a potent and selective 5-HT4 receptor (5-HT4R) partial agonist with a pKi of 8.7 in guinea-pig striatum. RS 67333 hydrochloride exhibits lower affinities at several other receptors including 5-HT1A 5-HT1D 5-HT2A 5-HT2C dopamine D1 D2 and muscarinic M1-M3 receptors. RS 67333 hydrochloride shows neuroprotective effects and can be used for Alzheimer's disease research. purity: 98%