Learn More
American Radiolabeled Chemicals Inc RS 79948 197 3H 50UC
SDP
Catalog No. 50901220
Encompass_Preferred
RS-79948-197-3H 50uCi in ethanol, 1 mCi/ml, S.A. 60-90 Ci/mmol, M.W. 364.5, Shipped in Dry Ice, Exclusive
Catalog No. 50-901-220 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 179250UC
RS-79948-197-3H 50uCi in ethanol, 1 mCi/ml, S.A. 60-90 Ci/mmol, M.W. 364.5, Shipped in Dry Ice, Exclusive