Learn More
Medchemexpress LLC RS-79948 hydrochloride hemihydrate | 186002-54-0 | 99.5% | 400.96 | C19H29ClN2O3S | 5 MG
SDP
Catalog No. 5000213204
Encompass_Preferred
RS-79948-197 hemihydrate is the hemihydrate salt of RS-79948, a selective non-imidazoline α2-adrenoceptor antagonist used in adrenergic receptor pharmacology research. Supplied as a white to off-white, high-purity solid in milligram quantities, it is intended for ligand binding, receptor characterization, and related in vitro or in vivo studies.
  • High purity (99.5%) suitable for sensitive assays.
  • Hemihydrate solid form for stable handling and storage.
  • Selective α2-adrenoceptor antagonist activity reported across species.
  • Small mg packaging appropriate for tracer and ligand studies.
  • Certificate of analysis and safety data available for each batch.
Catalog No. 50-002-13204 Supplier Medchemexpress LLC Supplier No. HY101338A5MG
RS-79948-197 hemihydrate is the hemihydrate salt of RS-79948, a selective non-imidazoline α2-adrenoceptor antagonist used in adrenergic receptor pharmacology research. Supplied as a white to off-white, high-purity solid in milligram quantities, it is intended for ligand binding, receptor characterization, and related in vitro or in vivo studies.
  • High purity (99.5%) suitable for sensitive assays.
  • Hemihydrate solid form for stable handling and storage.
  • Selective α2-adrenoceptor antagonist activity reported across species.
  • Small mg packaging appropriate for tracer and ligand studies.
  • Certificate of analysis and safety data available for each batch.