Learn More
TARGETMOL CHEMICALS INC RS 8359 5MG
SDP
Catalog No. 502581150
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RS 8359 is a reversible inhibitor of monoamine oxidase A (MAO-A) with a selectivity ratio of about 2200 for the AB enzyme types. RS 8359 has antidepressant activity. purity: 96%
Catalog No. 50-258-1150 Supplier TARGETMOL CHEMICALS INC Supplier No. T168035MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RS 8359 is a reversible inhibitor of monoamine oxidase A (MAO-A) with a selectivity ratio of about 2200 for the AB enzyme types. RS 8359 has antidepressant activity. purity: 96%