Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (RS)-AMPA hydrobromide | 171259-81-7 | 99.5% | 267.08 g/mol | C7H11BrN2O4 | 10 MG
SDP

Catalog No. 5000211964
Encompass_Preferred

(RS)-AMPA hydrobromide is the hydrobromide salt of (RS)-AMPA, a glutamate analogue and selective agonist of AMPA-type glutamate receptors. Supplied as a white to off-white solid for research use in neuroscience and pharmacology studies where a water-soluble AMPA receptor agonist is required.

  • Selective AMPA receptor agonist.
  • CAS number 171259-81-7.
  • Molecular weight 267.08 g/mol.
  • Purity 99.5%.
  • White to off-white solid.
  • Solubility: DMSO 50 mg/mL; H2O 5 mg/mL (may require ultrasonic).
  • Storage: sealed at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-11964 Supplier Medchemexpress LLC Supplier No. HY100815C10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(RS)-AMPA hydrobromide is the hydrobromide salt of (RS)-AMPA, a glutamate analogue and selective agonist of AMPA-type glutamate receptors. Supplied as a white to off-white solid for research use in neuroscience and pharmacology studies where a water-soluble AMPA receptor agonist is required.

  • Selective AMPA receptor agonist.
  • CAS number 171259-81-7.
  • Molecular weight 267.08 g/mol.
  • Purity 99.5%.
  • White to off-white solid.
  • Solubility: DMSO 50 mg/mL; H2O 5 mg/mL (may require ultrasonic).
  • Storage: sealed at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.