Learn More
Medchemexpress LLC (RS)-AMPA hydrobromide | 171259-81-7 | MFCD00055183 | 99.5% | 267.08 g/mol | C7H11BrN2O4 | 1 MG
SDP
Catalog No. 5000212315
Encompass_Preferred
(RS)-AMPA hydrobromide is the racemic hydrobromide salt of AMPA, a selective agonist of AMPA-type ionotropic glutamate receptors. It is intended for research use in receptor pharmacology, electrophysiology, and biochemical assays that require a water-soluble AMPA agonist.
  • Selective AMPA receptor agonist for glutamatergic signaling studies.
  • Hydrobromide salt form improves water solubility.
  • High purity suitable for analytical and biological experiments.
  • Available in small milligram pack sizes for laboratory use.
  • Applicable to electrophysiology, pharmacology, and biochemical assays.
Catalog No. 50-002-12315 Supplier Medchemexpress LLC Supplier No. HY100815C1MG
(RS)-AMPA hydrobromide is the racemic hydrobromide salt of AMPA, a selective agonist of AMPA-type ionotropic glutamate receptors. It is intended for research use in receptor pharmacology, electrophysiology, and biochemical assays that require a water-soluble AMPA agonist.
  • Selective AMPA receptor agonist for glutamatergic signaling studies.
  • Hydrobromide salt form improves water solubility.
  • High purity suitable for analytical and biological experiments.
  • Available in small milligram pack sizes for laboratory use.
  • Applicable to electrophysiology, pharmacology, and biochemical assays.