Learn More
Medchemexpress LLC (RS)-AMPA hydrobromide | 171259-81-7 | MFCD00055183 | 99.5% | 267.08 g·mol⁻¹ | C7H11BrN2O4 | 5 MG
SDP
Catalog No. 5000212334
Encompass_Preferred
(RS)-AMPA hydrobromide is a research-grade glutamate analogue that acts as a potent, selective agonist of AMPA-type ionotropic glutamate receptors. Supplied as a solid hydrobromide salt with high purity, it is intended for use in neuropharmacology, electrophysiology, and receptor pharmacology studies. The compound is soluble in DMSO and water (sonication recommended) and should be stored protected from moisture under refrigerated or low-temperature conditions as specified by the supplier.
  • Potent, selective agonist of AMPA receptors.
  • Hydrobromide salt increases aqueous solubility.
  • High purity suitable for research and analytical applications.
  • Soluble in DMSO and water; sonication recommended.
  • Stable when stored sealed and protected from moisture.
  • Provided in small milligram quantities for experimental use.
Catalog No. 50-002-12334 Supplier Medchemexpress LLC Supplier No. HY100815C5MG
(RS)-AMPA hydrobromide is a research-grade glutamate analogue that acts as a potent, selective agonist of AMPA-type ionotropic glutamate receptors. Supplied as a solid hydrobromide salt with high purity, it is intended for use in neuropharmacology, electrophysiology, and receptor pharmacology studies. The compound is soluble in DMSO and water (sonication recommended) and should be stored protected from moisture under refrigerated or low-temperature conditions as specified by the supplier.
  • Potent, selective agonist of AMPA receptors.
  • Hydrobromide salt increases aqueous solubility.
  • High purity suitable for research and analytical applications.
  • Soluble in DMSO and water; sonication recommended.
  • Stable when stored sealed and protected from moisture.
  • Provided in small milligram quantities for experimental use.