Learn More
Medchemexpress LLC (RS)-AMPA monohydrate | 76463-67-7 | 99.9% | 204.18 g/mol | C7H12N2O5 | 1 MG
SDP
Catalog No. 5000212335
Encompass_Preferred
(RS)-AMPA monohydrate is the racemic monohydrate form of AMPA, a glutamate analogue and selective agonist of AMPA-type ionotropic glutamate receptors. Supplied as a solid for research use in neuropharmacology and receptor pharmacology, it is used to investigate excitatory neurotransmission, receptor activation, and signaling pathways.
  • Racemic monohydrate form suitable for receptor pharmacology studies.
  • High purity for reproducible experimental results.
  • Solid form convenient for accurate dosing and storage.
  • Available in small pack sizes to support low-volume experiments.
  • Well characterized with defined molecular weight and CAS identifier for reliable identification.
Catalog No. 50-002-12335 Supplier Medchemexpress LLC Supplier No. HY100815D1MG
(RS)-AMPA monohydrate is the racemic monohydrate form of AMPA, a glutamate analogue and selective agonist of AMPA-type ionotropic glutamate receptors. Supplied as a solid for research use in neuropharmacology and receptor pharmacology, it is used to investigate excitatory neurotransmission, receptor activation, and signaling pathways.
  • Racemic monohydrate form suitable for receptor pharmacology studies.
  • High purity for reproducible experimental results.
  • Solid form convenient for accurate dosing and storage.
  • Available in small pack sizes to support low-volume experiments.
  • Well characterized with defined molecular weight and CAS identifier for reliable identification.