Learn More
Apexbio Technology LLC R-S-ATENOLOL-5G
SDP
Catalog No. 502240600
Encompass_Preferred
(R S)-Atenolol (CAS No 29122-68-7) is a beta-adrenergic receptor antagonist commonly used in cardiovascular research due to its effects on heart rate and blood pressure Originating from the need to manage hypertension and angina its mechanism of action involves blocking beta-1 adrenergic receptors which decreases cardiac output and suppresses renin release In vitro activity is observed in the nanomolar to low micromolar range making it effective in studies on cardiovascular cells and tissue cultures Typical applications include investigations in various cell models like cardiomyocytes and animal studies to evaluate efficacy and pharmacokinetics It is also utilized in drug screening for cardiovascular therapies Experimental concentrations typically depend on the specific research design
This item is not returnable. View return policy
Catalog No. 50-224-0600 Supplier Apexbio Technology LLC Supplier No. C52865000
(R S)-Atenolol (CAS No 29122-68-7) is a beta-adrenergic receptor antagonist commonly used in cardiovascular research due to its effects on heart rate and blood pressure Originating from the need to manage hypertension and angina its mechanism of action involves blocking beta-1 adrenergic receptors which decreases cardiac output and suppresses renin release In vitro activity is observed in the nanomolar to low micromolar range making it effective in studies on cardiovascular cells and tissue cultures Typical applications include investigations in various cell models like cardiomyocytes and animal studies to evaluate efficacy and pharmacokinetics It is also utilized in drug screening for cardiovascular therapies Experimental concentrations typically depend on the specific research design