Learn More
Medchemexpress LLC (RS)-CPP | 100828-16-8 | MFCD00055136 | 99.0% | 252.20 g/mol | C8H17N2O5P | 1 MG
SDP
Catalog No. 5000212542
Encompass_Preferred
(RS)-CPP is a research-grade NMDA receptor antagonist used to probe NMDA-mediated neurotransmission and excitotoxicity in in vitro and in vivo studies. It inhibits central neuron responses and has reported anticonvulsant activity.
  • Potent and selective NMDA receptor antagonist.
  • Suitable for electrophysiology, pharmacology, and anticonvulsant studies.
  • High purity (99.0%) for reproducible experimental results.
  • Water-soluble; may require sonication for complete dissolution.
  • Stable as a powder when stored frozen to preserve shelf life.
Catalog No. 50-002-12542 Supplier Medchemexpress LLC Supplier No. HY1007971MG
(RS)-CPP is a research-grade NMDA receptor antagonist used to probe NMDA-mediated neurotransmission and excitotoxicity in in vitro and in vivo studies. It inhibits central neuron responses and has reported anticonvulsant activity.
  • Potent and selective NMDA receptor antagonist.
  • Suitable for electrophysiology, pharmacology, and anticonvulsant studies.
  • High purity (99.0%) for reproducible experimental results.
  • Water-soluble; may require sonication for complete dissolution.
  • Stable as a powder when stored frozen to preserve shelf life.