Learn More
Abcam (R,S)-Rolipram, Selective PDE4 inhibitor, 50MG
Catalog No. 502707917
Encompass_Preferred
MW 275.34 Da, Purity >99%. Selective cAMP-specific phosphodiesterase PDE4 inhibitor (IC₅₀ values are 1.1 and 0.17 μM for inhibition of mouse PDE4A1 and PDE4B, respectively).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7917 Supplier Abcam Supplier No. AB12002950MG
MW 275.34 Da, Purity >99%. Selective cAMP-specific phosphodiesterase PDE4 inhibitor (IC₅₀ values are 1.1 and 0.17 μM for inhibition of mouse PDE4A1 and PDE4B, respectively).
The product is subject to the following: Abcam Restricted Use Statement