Learn More
Medchemexpress LLC Rs100329 hydrochloride | 1215654-26-4 | 99.6% | 462.89 g/mol | C20H26ClF3N4O3 | 10 MG
SDP
Catalog No. 5000247720
Encompass_Preferred
RS100329 hydrochloride is a selective α1A-adrenoceptor antagonist supplied as the hydrochloride salt for research use. It is reported to have high affinity for α1A (pKi ≈ 9.6) with lower affinity for α1D and α1B, and is used in preclinical studies including investigations of urethral reflex contractions and benign prostatic hyperplasia.
  • Selective α1A-adrenoceptor antagonist for receptor pharmacology research.
  • High reported purity suitable for preclinical assays.
  • Supplied as a stable hydrochloride salt in solid form.
  • Available in small milligram pack sizes for early-stage studies.
Catalog No. 50-002-47720 Supplier Medchemexpress LLC Supplier No. HY10320410MG
RS100329 hydrochloride is a selective α1A-adrenoceptor antagonist supplied as the hydrochloride salt for research use. It is reported to have high affinity for α1A (pKi ≈ 9.6) with lower affinity for α1D and α1B, and is used in preclinical studies including investigations of urethral reflex contractions and benign prostatic hyperplasia.
  • Selective α1A-adrenoceptor antagonist for receptor pharmacology research.
  • High reported purity suitable for preclinical assays.
  • Supplied as a stable hydrochloride salt in solid form.
  • Available in small milligram pack sizes for early-stage studies.