Learn More
Medchemexpress LLC RS100329 hydrochloride | 1215654-26-4 | 99.6% | 462.89 g/mol | C20H26ClF3N4O3 | 100 MG
SDP
Catalog No. 5000247453
Encompass_Preferred
RS100329 hydrochloride is the hydrochloride salt of a potent and subtype-selective α1A-adrenoceptor antagonist used as a research reagent in receptor pharmacology and selectivity studies. The compound is supplied as a solid, with a reported molecular weight of 462.89 g/mol and high batch-specific purity; consult the certificate of analysis for exact values and handling instructions.
  • Potent, selective α1A-adrenoceptor antagonist.
  • Hydrochloride salt form for improved stability and handling.
  • High purity suitable for in vitro pharmacology (COA available).
  • Supplied as a solid for dissolution in common organic solvents.
  • Used in receptor binding and functional assays to assess selectivity.
Catalog No. 50-002-47453 Supplier Medchemexpress LLC Supplier No. HY103204100MG
RS100329 hydrochloride is the hydrochloride salt of a potent and subtype-selective α1A-adrenoceptor antagonist used as a research reagent in receptor pharmacology and selectivity studies. The compound is supplied as a solid, with a reported molecular weight of 462.89 g/mol and high batch-specific purity; consult the certificate of analysis for exact values and handling instructions.
  • Potent, selective α1A-adrenoceptor antagonist.
  • Hydrochloride salt form for improved stability and handling.
  • High purity suitable for in vitro pharmacology (COA available).
  • Supplied as a solid for dissolution in common organic solvents.
  • Used in receptor binding and functional assays to assess selectivity.