Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RS100329 hydrochloride | 1215654-26-4 | 99.6% | 462.89 g/mol | C20H26ClF3N4O3 | 25 MG
SDP

Catalog No. 5000247451
Encompass_Preferred

RS100329 hydrochloride is the hydrochloride salt of RS100329, a potent and selective α1A-adrenoceptor antagonist used in pharmacological research, including studies of urethral reflex contractions and benign prostatic hyperplasia. It is supplied as a solid research chemical with high reported purity and defined storage recommendations.

  • Potent, selective α1A-adrenoceptor antagonist (pKi 9.6 for α1A).
  • High purity (≈99.6% by HPLC), suitable for biochemical assays.
  • Well-characterized small molecule: molecular weight 462.89 g/mol, formula C20H26ClF3N4O3.
  • Provided as a solid hydrochloride salt for accurate dosing in experiments.
  • Recommended storage: 4°C protected from light; in solvent, -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-47451 Supplier Medchemexpress LLC Supplier No. HY10320425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RS100329 hydrochloride is the hydrochloride salt of RS100329, a potent and selective α1A-adrenoceptor antagonist used in pharmacological research, including studies of urethral reflex contractions and benign prostatic hyperplasia. It is supplied as a solid research chemical with high reported purity and defined storage recommendations.

  • Potent, selective α1A-adrenoceptor antagonist (pKi 9.6 for α1A).
  • High purity (≈99.6% by HPLC), suitable for biochemical assays.
  • Well-characterized small molecule: molecular weight 462.89 g/mol, formula C20H26ClF3N4O3.
  • Provided as a solid hydrochloride salt for accurate dosing in experiments.
  • Recommended storage: 4°C protected from light; in solvent, -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.