Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RS100329 hydrochloride | 1215654-26-4 | 99.6% | 462.89 g/mol | C20H26ClF3N4O3 | 5 MG
SDP

Catalog No. 5000247719
Encompass_Preferred

RS100329 hydrochloride is a selective α1A-adrenoceptor antagonist for research use that inhibits reflex urethral contractions and is used in studies of benign prostatic hyperplasia. The compound is supplied as the hydrochloride salt with high analytical purity and characterized molecular properties.

  • CAS number 1215654-26-4.
  • Molecular formula C20H26ClF3N4O3.
  • Molecular weight 462.89 g/mol.
  • Purity 99.6%.
  • Solubility in DMSO 31.25 mg/mL (67.51 mM); ultrasonic assistance recommended.
  • Recommended storage solid at 4°C, protect from light; in solvent -80°C for up to 6 months, -20°C for up to 1 month.
  • Intended for research use only; not for human or veterinary use.

Catalog No. 50-002-47719 Supplier Medchemexpress LLC Supplier No. HY1032045MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RS100329 hydrochloride is a selective α1A-adrenoceptor antagonist for research use that inhibits reflex urethral contractions and is used in studies of benign prostatic hyperplasia. The compound is supplied as the hydrochloride salt with high analytical purity and characterized molecular properties.

  • CAS number 1215654-26-4.
  • Molecular formula C20H26ClF3N4O3.
  • Molecular weight 462.89 g/mol.
  • Purity 99.6%.
  • Solubility in DMSO 31.25 mg/mL (67.51 mM); ultrasonic assistance recommended.
  • Recommended storage solid at 4°C, protect from light; in solvent -80°C for up to 6 months, -20°C for up to 1 month.
  • Intended for research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.