Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RS100329 hydrochloride | 1215654-26-4 | MFCD00953081 | 99.6% | 462.89 g·mol⁻1 | C20H26ClF3N4O3 | 50 MG
SDP

Catalog No. 5000246875
Encompass_Preferred

RS100329 hydrochloride is the hydrochloride salt of RS100329, a potent and selective α1A-adrenoceptor antagonist used in pharmacological research. It is supplied as a white to off-white solid with high reported purity, and is suitable for in vitro and in vivo studies of α1A-adrenoceptor signaling and pharmacology.

  • High purity (99.57%), white to off-white solid.
  • Selective α1A-adrenoceptor antagonist for receptor pharmacology studies.
  • Suitable for in vitro and in vivo research applications.
  • Molecular weight 462.89 g·mol⁻1; formula C20H26ClF3N4O3.
  • Store at 4°C and protect from light; solutions: -80°C (6 months) or -20°C (1 month).
  • Supplied in 50 mg quantities.

Catalog No. 50-002-46875 Supplier Medchemexpress LLC Supplier No. HY10320450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RS100329 hydrochloride is the hydrochloride salt of RS100329, a potent and selective α1A-adrenoceptor antagonist used in pharmacological research. It is supplied as a white to off-white solid with high reported purity, and is suitable for in vitro and in vivo studies of α1A-adrenoceptor signaling and pharmacology.

  • High purity (99.57%), white to off-white solid.
  • Selective α1A-adrenoceptor antagonist for receptor pharmacology studies.
  • Suitable for in vitro and in vivo research applications.
  • Molecular weight 462.89 g·mol⁻1; formula C20H26ClF3N4O3.
  • Store at 4°C and protect from light; solutions: -80°C (6 months) or -20°C (1 month).
  • Supplied in 50 mg quantities.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.