Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RS100329 hydrochloride (2,4(1H,3H)-pyrimidinedione derivative) | 1215654-26-4 | 99.6% | 462.89 | C20H26ClF3N4O3 | 1 ML
SDP

Catalog No. 5000247270
Encompass_Preferred

RS100329 hydrochloride is a potent, selective α1A-adrenoceptor antagonist provided for in vitro receptor pharmacology and biochemical assays. It is available as a pre-made 10 mM solution in DMSO (1 mL) and in multiple solid sizes, with accompanying analytical documentation for research use.

  • Potent, selective α1A-adrenoceptor antagonist suitable for in vitro studies.
  • Available as 10 mM DMSO solution (1 mL) and as solid in multiple mg sizes.
  • High purity (99.57%) appropriate for biochemical assays.
  • Molecular weight 462.89 and formula C20H26ClF3N4O3.
  • Supplied with data sheet, certificate of analysis, and safety data sheet.
  • Intended for research use; follow laboratory safety and SDS guidance.

Catalog No. 50-002-47270 Supplier Medchemexpress LLC Supplier No. HY10320410MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RS100329 hydrochloride is a potent, selective α1A-adrenoceptor antagonist provided for in vitro receptor pharmacology and biochemical assays. It is available as a pre-made 10 mM solution in DMSO (1 mL) and in multiple solid sizes, with accompanying analytical documentation for research use.

  • Potent, selective α1A-adrenoceptor antagonist suitable for in vitro studies.
  • Available as 10 mM DMSO solution (1 mL) and as solid in multiple mg sizes.
  • High purity (99.57%) appropriate for biochemical assays.
  • Molecular weight 462.89 and formula C20H26ClF3N4O3.
  • Supplied with data sheet, certificate of analysis, and safety data sheet.
  • Intended for research use; follow laboratory safety and SDS guidance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.