Learn More
ATCC RS4 (11); Leukemia; Human (Homo sapiens)
Catalog No. 502384264
Encompass
RS4;11 is a lymphoblast cell line that was isolated from the marrow of a White, 32-year-old, female patient with acute lymphoblastic leukemia. This cell line was deposited by JH Kersey. It can be used in immunology and immune system disorder research.
Catalog No. 50-238-4264 Supplier ATCC Supplier No. CRL1873
RS4;11 is a lymphoblast cell line that was isolated from the marrow of a White, 32-year-old, female patient with acute lymphoblastic leukemia. This cell line was deposited by JH Kersey. It can be used in immunology and immune system disorder research.