Learn More
Roboz Surgical RS6470/MICRO VAS CLIP
SDP
Catalog No. NC9833142
Encompass
Micro Vascular Clip; 1mm Vessel Diameter; 30-35 Grams Occlusion Pressure; 8mm Length. Ideal for clamping of small animal vessels.
Catalog No. NC9833142 Supplier Roboz Surgical Supplier No. RS6470
Micro Vascular Clip; 1mm Vessel Diameter; 30-35 Grams Occlusion Pressure; 8mm Length. Ideal for clamping of small animal vessels.